Tisk…

Anti-TFB1M Goat Polyclonal Antibody

Dodavatel: Abnova
PAB16639
ABNOPAB16639EA 14910 CZK
ABNOPAB16639
Anti-TFB1M Goat Polyclonal Antibody
Protilátky
Goat polyclonal antibody raised against synthetic peptide of TFB1M.

Recommended Dilutions: ELISA: 1:64000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: TFB1M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR