Tisk…

Anti-IgG Rabbit Monoclonal Antibody (Texas Red®)

Dodavatel: Abnova
PAB10779
ABNOPAB10779EA 12060 CZK
ABNOPAB10779
Anti-IgG Rabbit Monoclonal Antibody (Texas Red®)
Protilátky
Rabbit anti-mouse IgG recognizes mouse IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red.

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Texas Red®
Epitope: F(c)
Host: Rabbit
Isotype:
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR