Tisk…
BSA I Restriction Endonuclease
Dodavatel: Abnova
P9388.250UL P9387.50UL
ABNOP9388.250ULEA 20270 CZK
ABNOP9388.250UL ABNOP9387.50UL
BSA I Restriction Endonuclease
Činidla pro nukleové kyseliny Restriction Enzymes
The BSA I restriction endonuclease is a restriction endonuclease BsaI that can recognize specific sites expressed by Escherichia coli recombinantly. Restriction endonucleases are widely used in various fields such as gene positioning and cloning. This enzyme cleaves DNA rapidly for efficient gene linearization.
Objednat
Produktová specifikace
Environmentally Preferable
Aplikace
Název enzymu BSA I
Zdroj