Tisk…

Anti-CLU Mouse Monoclonal Antibody [clone: CLI-9]

Dodavatel: Abnova
MAB3482
ABNOMAB3482EA 33350 CZK
ABNOMAB3482
Anti-CLU Mouse Monoclonal Antibody [clone: CLI-9]
Protilátky
Mouse monoclonal antibody raised against CLU.

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: CLU
Clonality: Monoclonal
Clone: CLI-9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR