Tisk…

Anti-Csf1r Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: CSF-1R]

Dodavatel: Abnova
MAB15526
ABNOMAB15526EA 12060 CZK
ABNOMAB15526
Anti-Csf1r Rat Monoclonal Antibody (PerCP-Cyanine5.5) [clone: CSF-1R]
Protilátky
Rat monoclonal antibody raised against recombinant mouse Csf1r.

Type: Primary
Antigen: Csf1r
Clonality: Monoclonal
Clone: CSF-1R
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR