Tisk…

Anti-Cd45.1 Mouse Monoclonal Antibody (Biotin) [clone: A20]

Dodavatel: Abnova
MAB15507
ABNOMAB15507EA 11460 CZK
ABNOMAB15507
Anti-Cd45.1 Mouse Monoclonal Antibody (Biotin) [clone: A20]
Protilátky
Mouse monoclonal antibody raised against native mouse Cd45.1.

Type: Primary
Antigen: Cd45.1
Clonality: Monoclonal
Clone: A20
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR