Tisk…

Anti-RBP Mouse Monoclonal Antibody [clone: SPM442]

Dodavatel: Abnova
MAB15211
ABNOMAB15211EA 14910 CZK
ABNOMAB15211
Anti-RBP Mouse Monoclonal Antibody [clone: SPM442]
Protilátky
Mouse monoclonal antibody raised against native human Retinol Binding Protein.

Type: Primary
Antigen: RBP
Clonality: Monoclonal
Clone: SPM442
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR