Tisk…

Anti-ACPP Mouse Monoclonal Antibody [clone: ACPP/1338]

Dodavatel: Abnova
MAB14881
ABNOMAB14881EA 14910 CZK
ABNOMAB14881
Anti-ACPP Mouse Monoclonal Antibody [clone: ACPP/1338]
Protilátky
Mouse monoclonal antibody raised against full length recombinant human ACPP.

Type: Primary
Antigen: ACPP
Clonality: Monoclonal
Clone: ACPP/1338
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR