Tisk…

Anti-dsDNA Mouse Monoclonal Antibody [clone: DSD/958]

Dodavatel: Abnova
MAB14636
ABNOMAB14636EA 14910 CZK
ABNOMAB14636
Anti-dsDNA Mouse Monoclonal Antibody [clone: DSD/958]
Protilátky
Mouse monoclonal antibody raised against human double stranded DNA.

Type: Primary
Antigen: dsDNA
Clonality: Monoclonal
Clone: DSD/958
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3, kappa
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu dsDNA
Environmentally Preferable
Druh protilátky Primární
Klonovalita Monoclonal
Klon DSD/958
Reaktivita Human
Hostitel Mouse
Izotyp IgG3 kappa
Imunochemie Yes
Imunogen Nuclei of Burkitt's cells.
Formulář Liquid
Čištění Protein G purification
Skladovací pufr In 10 mM PBS.
Skladovací teplota Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR