Tisk…

Anti-GJB1 Mouse Monoclonal Antibody [clone: GJB1/1753]

Dodavatel: Abnova
MAB14546
ABNOMAB14546EA 14910 CZK
ABNOMAB14546
Anti-GJB1 Mouse Monoclonal Antibody [clone: GJB1/1753]
Protilátky
Mouse monoclonal antibody raised against full length recombinant human GJB1.

Type: Primary
Antigen: GJB1
Clonality: Monoclonal
Clone: GJB1/1753
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR