Tisk…

Anti-ENO2 Mouse Monoclonal Antibody [clone: ENO2/1462]

Dodavatel: Abnova
MAB14516
ABNOMAB14516EA 14910 CZK
ABNOMAB14516
Anti-ENO2 Mouse Monoclonal Antibody [clone: ENO2/1462]
Protilátky
Mouse monoclonal antibody raised against synthetic peptide of human ENO2.

Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: ENO2/1462
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse;Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR