Tisk…

Anti-ADFP Mouse Monoclonal Antibody [clone: ADFP/1493]

Dodavatel: Abnova
MAB14486
ABNOMAB14486EA 14910 CZK
ABNOMAB14486
Anti-ADFP Mouse Monoclonal Antibody [clone: ADFP/1493]
Protilátky
Mouse monoclonal antibody raised against partial recombinant human ADFP.

Type: Primary
Antigen: ADFP
Clonality: Monoclonal
Clone: ADFP/1493
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, lambda
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR