Tisk…

Anti-MCM9 Rabbit Polyclonal Antibody

Dodavatel: Abnova
H00254394-D01P
ABNOH00254394-D01PEA 10380 CZK
ABNOH00254394-D01P
Anti-MCM9 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MCM9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR