Tisk…

Anti-AMOT Polyclonal Antibody Pair

Dodavatel: Abnova
H00154796-AP21
ABNOH00154796-AP21EA 24000 CZK
ABNOH00154796-AP21
Anti-AMOT Polyclonal Antibody Pair
Protilátky
This Set comes with matched Antibody Pair to detect and quantify protein level of human AMOT.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-AMOT (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-AMOT (20 µg);

Type:
Antigen: AMOT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu AMOT
Název antigenu Angiomotin
Environmentally Preferable
Klonovalita Polyclonal
Reaktivita Human
Hostitel Multiple
Identifikační číslo genu 154796
Izotyp
Skladovací teplota Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR