Tisk…

Anti-RAB1B Mouse Monoclonal Antibody [clone: 1B2]

Dodavatel: Abnova
H00081876-M02
ABNOH00081876-M02EA 11320 CZK
ABNOH00081876-M02
Anti-RAB1B Mouse Monoclonal Antibody [clone: 1B2]
Protilátky
Type: Primary
Antigen: RAB1B
Clonality: Monoclonal
Clone: 1B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR