Tisk…

Anti-ZAK Mouse Monoclonal Antibody [clone: 3G5]

Dodavatel: Abnova
H00051776-M03A
ABNOH00051776-M03AEA 11320 CZK
ABNOH00051776-M03A
Anti-ZAK Mouse Monoclonal Antibody [clone: 3G5]
Protilátky
Type: Primary
Antigen: ZAK
Clonality: Monoclonal
Clone: 3G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR