Tisk…

Anti-DLL1 Mouse Monoclonal Antibody [clone: 4F9]

Dodavatel: Abnova
H00028514-M02
ABNOH00028514-M02EA 11320 CZK
ABNOH00028514-M02
Anti-DLL1 Mouse Monoclonal Antibody [clone: 4F9]
Protilátky
Type: Primary
Antigen: DLL1
Clonality: Monoclonal
Clone: 4F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR