Tisk…

Anti-PCSK1N Mouse Monoclonal Antibody [clone: 1E9]

Dodavatel: Abnova
H00027344-M02
ABNOH00027344-M02EA 11320 CZK
ABNOH00027344-M02
Anti-PCSK1N Mouse Monoclonal Antibody [clone: 1E9]
Protilátky
Type: Primary
Antigen: PCSK1N
Clonality: Monoclonal
Clone: 1E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR