Tisk…

Anti-RGL1 Mouse Monoclonal Antibody [clone: 3E10]

Dodavatel: Abnova
H00023179-M02
ABNOH00023179-M02EA 11320 CZK
ABNOH00023179-M02
Anti-RGL1 Mouse Monoclonal Antibody [clone: 3E10]
Protilátky
Type: Primary
Antigen: RGL1
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR