Tisk…

Anti-USH1C Mouse Monoclonal Antibody [clone: 2B9]

Dodavatel: Abnova
H00010083-M04
ABNOH00010083-M04EA 11320 CZK
ABNOH00010083-M04
Anti-USH1C Mouse Monoclonal Antibody [clone: 2B9]
Protilátky
Type: Primary
Antigen: USH1C
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR