Tisk…

Anti-ADAMTS4 Mouse Monoclonal Antibody [clone: 2A6]

Dodavatel: Abnova
H00009507-M01
ABNOH00009507-M01EA 11320 CZK
ABNOH00009507-M01
Anti-ADAMTS4 Mouse Monoclonal Antibody [clone: 2A6]
Protilátky
Type: Primary
Antigen: ADAMTS4
Clonality: Monoclonal
Clone: 2A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR