Tisk…

Anti-HAND2 Mouse Monoclonal Antibody [clone: 4B11]

Dodavatel: Abnova
H00009464-M08
ABNOH00009464-M08EA 11320 CZK
ABNOH00009464-M08
Anti-HAND2 Mouse Monoclonal Antibody [clone: 4B11]
Protilátky
Type: Primary
Antigen: HAND2
Clonality: Monoclonal
Clone: 4B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR