Tisk…

Anti-RARRES2 Rabbit Polyclonal Antibody

Dodavatel: Abnova
H00005919-D01
ABNOH00005919-D01EA 10380 CZK
ABNOH00005919-D01
Anti-RARRES2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RARRES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR