Tisk…

Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: 6C6]

Dodavatel: Abnova
H00003115-M01
ABNOH00003115-M01EA 11320 CZK
ABNOH00003115-M01
Anti-HLA-DPB1 Mouse Monoclonal Antibody [clone: 6C6]
Protilátky
Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: 6C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR