Tisk…

Anti-CHAT Mouse Monoclonal Antibody [clone: 1H7]

Dodavatel: Abnova
H00001103-M01
ABNOH00001103-M01EA 11320 CZK
ABNOH00001103-M01
Anti-CHAT Mouse Monoclonal Antibody [clone: 1H7]
Protilátky
Type: Primary
Antigen: CHAT
Clonality: Monoclonal
Clone: 1H7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR