Tisk…

Anti-DNAJC6 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP9982A
ABGEAP9982AEA 10080 CZK
ABGEAP9982A
Anti-DNAJC6 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DNAJC6 (DnaJ (Hsp40) homolog, subfamily C, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Produktová specifikace

Symbol antigenu DNAJC6
Název antigenu DnaJ (Hsp40) homolog, subfamily C, member 6
Environmentally Preferable
Synonyma antigenu DJC6|PARK19
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Mouse
Hostitel Rabbit
Identifikační číslo genu 6923
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Průtoková cytometrie Yes
Epitop Center
Imunogen This TCEB2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 21-48 amino acids from the Central region of human TCEB2.
Sekvence KRPPDEQRLYKDDC
Čištění Protein A & Peptide affinity purification
Koncentrace 0,4 mg/ml
Skladovací pufr PBS with 0,09% (W/V) sodium azide, pH 7,4
Skladovací teplota Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR