Tisk…

Anti-GSTK1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP9867C
ABGEAP9867CEA 10080 CZK
ABGEAP9867C
Anti-GSTK1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: GSTK1 (glutathione S-transferase kappa 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Produktová specifikace

Symbol antigenu GSTK1
Název antigenu Glutathione S-transferase kappa 1
Environmentally Preferable
Synonyma antigenu MOR42-1|S50
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Mouse
Hostitel Rabbit
Identifikační číslo genu 258837
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Průtoková cytometrie Yes
Epitop C-Terminal
Imunogen This Mouse Olfr545 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 281-308 amino acids from the C-terminal region of mouse Olfr545.
Sekvence YGARTKEIRQHLVR
Čištění Protein A & Peptide affinity purification
Koncentrace 0,45 mg/ml
Skladovací pufr PBS with 0,09% (W/V) sodium azide, pH 7,4
Skladovací teplota Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Transportní teplota 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR