Tisk…

Anti-FOXP1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP9849B
ABGEAP9849BEA 10080 CZK
ABGEAP9849B
Anti-FOXP1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: FOXP1 (forkhead box P1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR