Tisk…

Anti-PLCB1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP9374B
ABGEAP9374BEA 10080 CZK
ABGEAP9374B
Anti-PLCB1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PLCB1 (phospholipase C, beta 1 (phosphoinositide-specific))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR