Tisk…

Anti-HLA-G Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP8941C
ABGEAP8941CEA 10080 CZK
ABGEAP8941C
Anti-HLA-G Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HLA-G (major histocompatibility complex, class I, G)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR