Tisk…

Anti-PCK2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP8094A
ABGEAP8094AEA 10080 CZK
ABGEAP8094A
Anti-PCK2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PCK2 (phosphoenolpyruvate carboxykinase 2 (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR