Tisk…

Anti-ESRRG Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP7870C
ABGEAP7870CEA 10080 CZK
ABGEAP7870C
Anti-ESRRG Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ESRRG (estrogen-related receptor gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR