Tisk…

Anti-ERK3 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP7502A
ABGEAP7502AEA 10080 CZK
ABGEAP7502A
Anti-ERK3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ERK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (707–721 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR