Tisk…

Anti-REA (PHB2) Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP7270A
ABGEAP7270AEA 10080 CZK
ABGEAP7270A
Anti-REA (PHB2) Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: REA (PHB2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR