Tisk…

Anti-AKT1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP7141J
ABGEAP7141JEA 10080 CZK
ABGEAP7141J
Anti-AKT1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AKT1 (v-akt murine thymoma viral oncogene homolog 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR