Tisk…

Anti-EEF1A1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP6592B
ABGEAP6592BEA 10080 CZK
ABGEAP6592B
Anti-EEF1A1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: EEF1A1 (eukaryotic translation elongation factor 1 alpha 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR