Tisk…

Anti-JAG2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP6148A
ABGEAP6148AEA 10080 CZK
ABGEAP6148A
Anti-JAG2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: JAG2 (jagged 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR