Tisk…

Anti-CLDN12 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP5874A
ABGEAP5874AEA 10080 CZK
ABGEAP5874A
Anti-CLDN12 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CLDN12 (claudin 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR