Tisk…

Anti-ALG10B Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP5630B
ABGEAP5630BEA 10080 CZK
ABGEAP5630B
Anti-ALG10B Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ALG10B (ALG10B, alpha-1,2-glucosyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR