Tisk…

Anti-TSPAN3 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP5559C
ABGEAP5559CEA 10080 CZK
ABGEAP5559C
Anti-TSPAN3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TSPAN3 (tetraspanin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR