Tisk…

Anti-TTC26 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP5325B
ABGEAP5325BEA 10080 CZK
ABGEAP5325B
Anti-TTC26 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TTC26 (tetratricopeptide repeat domain 26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Produktová specifikace

Symbol antigenu TTC26
Environmentally Preferable
Název antigenu Tetratricopeptide Repeat Doman 26
Synonyma antigenu MLF1|Myelodysplasia-myeloid leukemia factor 1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human
Hostitel Rabbit
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Epitop C-terminal
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR