Tisk…

Anti-TOPBP1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP3774A
ABGEAP3774AEA 10080 CZK
ABGEAP3774A
Anti-TOPBP1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TOPBP1 (topoisomerase (DNA) II binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR