Tisk…

Anti-PI3KC3 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP3369A
ABGEAP3369AEA 10080 CZK
ABGEAP3369A
Anti-PI3KC3 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: PI3KC3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR