Tisk…

Anti-TNFRSF1A Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP3274A
ABGEAP3274AEA 10080 CZK
ABGEAP3274A
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: TNFRSF1A (tumor necrosis factor receptor superfamily, member 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Objednat

Produktová specifikace

Symbol antigenu TNFRSF1A
Environmentally Preferable
Název antigenu Tumor Necrosis Factor Receptor Superfamily Member 1A
Synonyma antigenu Tnfr1|p55-R|TNFRI|TNF-R|TNF-R-I|p55|CD120a|FPF|TNFAR|TNF-R1|Tnfr-2|TNFalpha-R1|TNFRp55|TNFR60|TNF-R55|TNF-alphaR1
Druh protilátky Primární
Klonovalita Polyclonal
Spojení Unconjugated
Reaktivita Human,Mouse
Hostitel Rabbit
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Křížová adsorbce No
Skladovací teplota Cool
Transportní teplota Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR