Tisk…

Anti-USP22 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP2148A
ABGEAP2148AEA 10080 CZK
ABGEAP2148A
Anti-USP22 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: USP22 (ubiquitin specific peptidase 22)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR