Tisk…

Anti-MAN2C1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP19732A
ABGEAP19732AEA 10080 CZK
ABGEAP19732A
Anti-MAN2C1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MAN2C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR