Tisk…

Anti-CHST2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP19294B
ABGEAP19294BEA 10080 CZK
ABGEAP19294B
Anti-CHST2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: CHST2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR