Tisk…

Anti-RGMA Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP19179B
ABGEAP19179BEA 10080 CZK
ABGEAP19179B
Anti-RGMA Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RGMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR