Tisk…

Anti-MCTP2 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP18655A
ABGEAP18655AEA 10080 CZK
ABGEAP18655A
Anti-MCTP2 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MCTP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR