Tisk…

Anti-OR56B1 Rabbit Polyclonal Antibody

Dodavatel: Abgent
AP18340A
ABGEAP18340AEA 10080 CZK
ABGEAP18340A
Anti-OR56B1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: OR56B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR